siRNA Details
Gene Symbol: PRPF40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017892.3 | NP_060362.3 | 6 | 382 | ||
XM_005246676.4 | XP_005246733.3 | 6 | 902 | |||
XM_005246677.4 | XP_005246734.2 | 6 | 378 | |||
XM_011511446.2 | XP_011509748.1 | 6 | 902 | |||
XM_011511447.2 | XP_011509749.1 | 6 | 902 | |||
XM_011511448.2 | XP_011509750.1 | 6 | 378 | |||
XM_011511449.2 | XP_011509751.1 | 8 | 732 | |||
XM_017004453.1 | XP_016859942.1 | 6 | 902 | |||
XM_017004454.1 | XP_016859943.1 | 6 | 902 | |||
XM_017004455.1 | XP_016859944.1 | 6 | 378 | |||
XM_017004456.1 | XP_016859945.1 | 7 | 568 | |||
XM_017004457.1 | XP_016859946.1 | 8 | 731 | |||
XM_017004458.1 | XP_016859947.1 | 7 | 532 | |||
XM_017004459.1 | XP_016859948.1 | 7 | 641 | |||
XM_017004460.1 | XP_016859949.1 | 7 | 569 | |||
XM_017004461.1 | XP_016859950.1 | 7 | 641 | |||
GenBank mRNA | AF049523.1 | - | 6 | 367 | ||
AF049524.1 | - | 6 | 320 | |||
AF049528.1 | - | 6 | 454 | |||
AK024810.1 | - | 7 | 701 | |||
BC000273.1 | - | 6 | 345 | |||
BC011788.2 | - | 6 | 366 | |||
BC029414.1 | - | 6 | 382 | |||
U70667.1 | - | 6 | 420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
