siRNA Details
Gene Symbol: PRPF40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017892.3 | NP_060362.3 | 12 | 1217 | ||
XM_005246676.4 | XP_005246733.3 | 12 | 1737 | |||
XM_005246677.4 | XP_005246734.2 | 12 | 1213 | |||
XM_011511446.2 | XP_011509748.1 | 11 | 1683 | |||
XM_011511447.2 | XP_011509749.1 | 12 | 1737 | |||
XM_011511448.2 | XP_011509750.1 | 11 | 1159 | |||
XM_011511449.2 | XP_011509751.1 | 14 | 1567 | |||
XM_017004453.1 | XP_016859942.1 | 12 | 1737 | |||
XM_017004454.1 | XP_016859943.1 | 11 | 1683 | |||
XM_017004455.1 | XP_016859944.1 | 11 | 1159 | |||
XM_017004456.1 | XP_016859945.1 | 13 | 1403 | |||
XM_017004457.1 | XP_016859946.1 | 14 | 1566 | |||
XM_017004458.1 | XP_016859947.1 | 12 | 1313 | |||
XM_017004459.1 | XP_016859948.1 | 12 | 1422 | |||
XM_017004460.1 | XP_016859949.1 | 12 | 1350 | |||
XM_017004461.1 | XP_016859950.1 | 12 | 1422 | |||
GenBank mRNA | AF049523.1 | - | 12 | 1202 | ||
AF049528.1 | - | 12 | 1289 | |||
AK024810.1 | - | 13 | 1536 | |||
AK298547.1 | - | 9 | 1054 | |||
BC000273.1 | - | 12 | 1180 | |||
BC011788.2 | - | 12 | 1201 | |||
BC029414.1 | - | 12 | 1217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
