siRNA Details
Gene Symbol: PRPF40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017892.3 | NP_060362.3 | 16 | 1763 | ||
XM_005246676.4 | XP_005246733.3 | 16 | 2283 | |||
XM_005246677.4 | XP_005246734.2 | 16 | 1759 | |||
XM_011511446.2 | XP_011509748.1 | 15 | 2229 | |||
XM_011511447.2 | XP_011509749.1 | 16 | 2283 | |||
XM_011511448.2 | XP_011509750.1 | 15 | 1705 | |||
XM_011511449.2 | XP_011509751.1 | 18 | 2113 | |||
XM_017004453.1 | XP_016859942.1 | 16 | 2283 | |||
XM_017004454.1 | XP_016859943.1 | 15 | 2229 | |||
XM_017004455.1 | XP_016859944.1 | 15 | 1705 | |||
XM_017004456.1 | XP_016859945.1 | 17 | 1949 | |||
XM_017004457.1 | XP_016859946.1 | 18 | 2112 | |||
XM_017004458.1 | XP_016859947.1 | 16 | 1859 | |||
XM_017004459.1 | XP_016859948.1 | 16 | 1968 | |||
XM_017004460.1 | XP_016859949.1 | 16 | 1896 | |||
XM_017004461.1 | XP_016859950.1 | 16 | 1968 | |||
GenBank mRNA | AF155096.1 | - | 2 | 200 | ||
AK021621.1 | - | 1 | 851 | |||
AK298547.1 | - | 13 | 1600 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
