siRNA Details
Gene Symbol: ZNF841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136499.1 | NP_001129971.1 | 7 | 3211 | ||
NM_001321349.1 | NP_001308278.1 | 5 | 2955 | |||
XM_011526833.2 | XP_011525135.1 | 6 | 3103 | |||
XM_017026652.1 | XP_016882141.1 | 3 | 2760 | |||
XM_017026653.1 | XP_016882142.1 | 7 | 3169 | |||
XM_017026654.1 | XP_016882143.1 | 7 | 3155 | |||
XM_017026655.1 | XP_016882144.1 | 3 | 2757 | |||
XM_017026656.1 | XP_016882145.1 | 6 | 4806 | |||
XM_017026657.1 | XP_016882146.1 | 6 | 3100 | |||
GenBank mRNA | AK093512.1 | - | 1 | 1692 | ||
AK094953.1 | - | 1 | 1316 | |||
AK129938.1 | - | 2 | 1514 | |||
AK131409.1 | - | 5 | 2955 | |||
BC136254.1 | - | 4 | 2801 | 9020241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
