siRNA Details
Gene Symbol: DCAF12L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013628.2 | NP_001013650.1 | 1 | 525 | ||
GenBank mRNA | BC136673.1 | - | 2 | 433 | 9020663 | |
BC136674.1 | - | 1 | 525 | 9020664 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
