siRNA Details
Gene Symbol: MVB12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033446.2 | NP_258257.1 | 10 | 3560 | ||
XM_005252297.1 | XP_005252354.1 | 10 | 3595 | |||
XM_017015278.1 | XP_016870767.1 | 9 | 3465 | |||
GenBank mRNA | AK000001.1 | - | 4 | 3245 | ||
AK024432.1 | - | 10 | 3555 | |||
AK024441.1 | - | 7 | 3082 | |||
BC000122.1 | - | 11 | 2049 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
