siRNA Details
Gene Symbol: C17orf97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013672.4 | NP_001013694.4 | 2 | 515 | ||
GenBank mRNA | AK128660.1 | - | 515 | |||
BC057385.1 | - | 2 | 466 | |||
BC070116.1 | - | 2 | 441 | |||
BC141806.1 | - | 2 | 514 | 40147362 | ||
KR780251.1 | - | 2 | 515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
