siRNA Details
Gene Symbol: SRGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170637.3 | NP_001164108.1 | 3 | 924 | ||
NM_001300952.1 | NP_001287881.1 | 3 | 924 | |||
NM_015326.4 | NP_056141.2 | 3 | 924 | |||
XM_005277510.2 | XP_005277567.1 | 4 | 1111 | |||
XM_005277511.2 | XP_005277568.1 | 4 | 1111 | |||
XM_005277514.1 | XP_005277571.1 | 4 | 1111 | |||
XM_005277515.1 | XP_005277572.1 | 4 | 1111 | |||
XM_011509354.1 | XP_011507656.1 | 4 | 1111 | |||
XM_011509355.1 | XP_011507657.1 | 4 | 1111 | |||
XM_011509356.2 | XP_011507658.1 | 4 | 1111 | |||
XM_011509359.1 | XP_011507661.1 | 4 | 1111 | |||
XM_017000839.1 | XP_016856328.1 | 2 | 785 | |||
XM_017000840.1 | XP_016856329.1 | 3 | 357 | |||
XM_017000841.1 | XP_016856330.1 | 4 | 1111 | |||
GenBank mRNA | AB007925.1 | - | 3 | 454 | ||
AK295845.1 | - | 3 | 680 | |||
BC132872.1 | - | 3 | 452 | 40146894 | ||
BC132874.1 | - | 3 | 452 | 40146896 | ||
BC144343.1 | - | 3 | 452 | 9052863 | ||
BC150646.1 | - | 3 | 877 |
Gene Symbol: SRGAP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271870.2 | NP_001258799.2 | 3 | 843 | ||
XM_017002085.1 | XP_016857574.1 | 3 | 922 | |||
XM_017002086.1 | XP_016857575.1 | 3 | 768 | |||
XM_017002087.1 | XP_016857576.1 | 3 | 922 | |||
XM_017002088.1 | XP_016857577.1 | 4 | 983 | |||
XM_017002089.1 | XP_016857578.1 | 5 | 1031 | |||
XM_017002090.1 | XP_016857579.1 | 4 | 983 | |||
XM_017002091.1 | XP_016857580.1 | 5 | 1031 | |||
GenBank mRNA | HY154277.1 | - | 2 | 170 |
Gene Symbol: SRGAP2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271872.1 | NP_001258801.1 | 4 | 987 | ||
XM_005277497.3 | XP_005277554.1 | 4 | 1082 | |||
XM_011542015.2 | XP_011540317.1 | 3 | 555 | |||
GenBank mRNA | BC112927.1 | - | 4 | 987 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
