siRNA Details
Gene Symbol: RAMP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024461.1 | 4 | 2044 | |||
NR_024462.1 | 5 | 757 | ||||
GenBank mRNA | AK057121.1 | - | 3 | 906 | ||
BC001607.1 | - | 4 | 1889 | |||
BC018638.2 | - | 5 | 757 | |||
BC033118.1 | - | 5 | 757 | |||
BC041021.1 | - | 4 | 1890 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
