siRNA Details
Gene Symbol: RAMP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024461.1 | 4 | 1726 | |||
GenBank mRNA | BC001607.1 | - | 4 | 1571 | ||
BC041021.1 | - | 4 | 1572 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
