siRNA Details
Gene Symbol: RAMP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024461.1 | 2 | 563 | |||
GenBank mRNA | AK057121.1 | - | 1 | 591 | ||
BC001607.1 | - | 2 | 409 | |||
BC034924.1 | - | 2 | 317 | |||
BC041021.1 | - | 2 | 410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
