siRNA Details
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007239.1 | NP_001007240.1 | 1 | 428 | ||
NM_015935.4 | NP_057019.3 | 1 | 428 | |||
XM_017001458.1 | XP_016856947.1 | 1 | 545 | |||
GenBank mRNA | AB020666.2 | - | 1 | 183 | ||
AB242174.1 | - | 1 | 353 | |||
AB242175.1 | - | 1 | 341 | |||
AF132936.1 | - | 1 | 353 | |||
AK027552.1 | - | 1 | 347 | |||
AK223493.1 | - | 1 | 389 | |||
AK291740.1 | - | 1 | 375 | |||
AK304471.1 | - | 2 | 209 | |||
AL049669.1 | - | 1 | 353 | |||
BC029083.1 | - | 1 | 182 | 4509086 | ||
BI091265.1 | - | 1 | 544 | |||
BM833930.1 | - | 1 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
