siRNA Details
Gene Symbol: TXNRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173513.1 | NP_001166984.1 | 11 | 1442 | ||
NM_052883.1 | NP_443115.1 | 11 | 1442 | |||
GenBank mRNA | AF133519.1 | - | 11 | 1110 | ||
AF171055.1 | - | 11 | 1116 | |||
AK299502.1 | - | 11 | 1419 | |||
BC030028.2 | - | 11 | 1420 | |||
BC050032.1 | - | 11 | 1429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
