siRNA Details
Gene Symbol: KIAA0125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026800.2 | 4 | 1120 | |||
GenBank mRNA | AF150199.1 | - | 4 | 430 | ||
AF161538.1 | - | 4 | 430 | |||
AK125627.1 | - | 4 | 1121 | |||
BC016766.1 | - | 4 | 442 | 4077490 | ||
BC030286.1 | - | 4 | 454 | 5223261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
