siRNA Details
Gene Symbol: LINC00342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057596.1 | - | 1 | 304 | ||
BC117383.1 | - | 1 | 274 | |||
BC126153.1 | - | 1 | 274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
