siRNA Details
Gene Symbol: SLC35F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029858.3 | NP_001025029.2 | 7 | 1108 | ||
XM_005266865.4 | XP_005266922.1 | 7 | 1408 | |||
GenBank mRNA | AK128717.1 | - | 5 | 456 | ||
AK295860.1 | - | 7 | 805 | |||
BC036493.1 | - | 7 | 876 | |||
BC114925.1 | - | 8 | 982 | 40025808 | ||
BX640761.1 | - | 4 | 369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
