siRNA Details
Gene Symbol: LOC100506990 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040092.1 | 6 | 2195 | |||
GenBank mRNA | AK094417.1 | - | 2 | 1565 | ||
AK095981.1 | - | 6 | 1845 | |||
AK129982.1 | - | 7 | 2331 | |||
AL137298.1 | - | 4 | 1286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
