siRNA Details
Gene Symbol: HYKK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013619.3 | NP_001013641.2 | 2 | 327 | ||
NM_001083612.1 | NP_001077081.1 | 2 | 327 | |||
XM_011521231.2 | XP_011519533.1 | 2 | 309 | |||
XM_017021922.1 | XP_016877411.1 | 2 | 309 | |||
GenBank mRNA | AK126453.1 | - | 2 | 309 | ||
BC132753.1 | - | 2 | 274 | 40146775 | ||
BC144383.1 | - | 2 | 274 | 9052904 | ||
BM045979.1 | - | 2 | 328 | |||
BM826406.1 | - | 1 | 16 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
