siRNA Details
Gene Symbol: KIF21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252100.1 | NP_001239029.1 | 2 | 517 | ||
NM_001252102.1 | NP_001239031.1 | 2 | 517 | |||
NM_001252103.1 | NP_001239032.1 | 2 | 517 | |||
NM_017596.3 | NP_060066.2 | 2 | 517 | |||
XM_017000731.1 | XP_016856220.1 | 2 | 328 | |||
GenBank mRNA | AB007918.2 | - | 2 | 517 | ||
AM177180.1 | - | 2 | 200 | |||
BC137281.1 | - | 2 | 225 | 9021278 | ||
BC144557.1 | - | 2 | 225 | 9053085 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
