siRNA Details
Gene Symbol: LOC439938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095259.1 | - | 1 | 1215 |
Gene Symbol: LCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130915.1 | 3 | 973 | |||
GenBank mRNA | AK023806.1 | - | 3 | 962 | ||
BC070061.1 | - | 3 | 973 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
