siRNA Details
Gene Symbol: LCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130915.1 | 3 | 667 | |||
GenBank mRNA | AK023806.1 | - | 3 | 656 | ||
BC070061.1 | - | 3 | 667 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
