siRNA Details
Gene Symbol: RAB40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005262083.4 | XP_005262140.1 | 2 | 6844 | ||
XM_011530872.2 | XP_011529174.1 | 2 | 6841 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
