siRNA Details
Gene Symbol: LOC283887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135196.1 | 1 | 509 | |||
NR_135197.1 | 1 | 509 | ||||
GenBank mRNA | AK093382.1 | - | 1 | 358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
