siRNA Details
Gene Symbol: LOC283887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135196.1 | 2 | 1077 | |||
NR_135197.1 | 2 | 1063 | ||||
GenBank mRNA | AK093382.1 | - | 2 | 926 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
