siRNA Details
Gene Symbol: SNHG17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015366.4 | 4 | 368 | |||
NR_027241.2 | 4 | 594 | ||||
GenBank mRNA | AK022745.1 | - | 2 | 1699 | ||
BC032119.2 | - | 4 | 560 | |||
BC052370.1 | - | 5 | 350 | |||
BM714185.1 | - | 4 | 473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
