siRNA Details
Gene Symbol: SNHG17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015366.4 | 7, 8 | 759 | |||
NR_027241.2 | 6, 7 | 872 | ||||
GenBank mRNA | AK022745.1 | - | 3, 4 | 1822 | ||
BC032119.2 | - | 6, 7 | 838 | |||
BC052370.1 | - | 6, 7 | 473 | |||
BX640887.1 | - | 5, 6 | 4878 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
