siRNA Details
Gene Symbol: ILF3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024333.1 | 1 | 1492 | |||
GenBank mRNA | AF439744.2 | - | 1 | 1102 | ||
AK075016.1 | - | 1 | 1456 | |||
AK311360.1 | - | 1 | 1460 | |||
AL832185.1 | - | 1 | 1466 | |||
BC044911.2 | - | 1 | 1467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
