siRNA Details
Gene Symbol: RNF216P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015449.1 | 6 | 981 | |||
NR_023384.1 | 8 | 1154 | ||||
NR_023385.1 | 5 | 866 | ||||
GenBank mRNA | AK124529.1 | - | 2 | 2359 | ||
AK303831.1 | - | 5 | 717 | |||
AK308526.1 | - | 5 | 801 | |||
AK309798.1 | - | 5 | 725 | |||
BC026736.1 | - | 6 | 860 | |||
BC068459.1 | - | 8 | 1123 | |||
BX538059.1 | - | 6 | 908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
