siRNA Details
Gene Symbol: RNF216P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015449.1 | 6 | 895 | |||
NR_023384.1 | 8 | 1068 | ||||
NR_023385.1 | 5 | 780 | ||||
GenBank mRNA | AK124529.1 | - | 2 | 2273 | ||
AK303831.1 | - | 5 | 631 | |||
AK308526.1 | - | 5 | 715 | |||
BC026736.1 | - | 6 | 774 | |||
BC068459.1 | - | 8 | 1037 | |||
BX538059.1 | - | 6 | 822 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
