siRNA Details
Gene Symbol: ZNF783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195220.1 | NP_001182149.1 | 6 | 2952 | ||
XM_011515701.2 | XP_011514003.1 | 6 | 2880 | |||
GenBank mRNA | AK022976.1 | - | 1 | 974 | ||
AL359620.1 | - | 1 | 1478 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
