siRNA Details
Gene Symbol: FAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008781.2 | NP_001008781.2 | 9 | 7983 | ||
XM_017017178.1 | XP_016872667.1 | 11 | 8168 | |||
XM_017017179.1 | XP_016872668.1 | 12 | 8390 | |||
XM_017017180.1 | XP_016872669.1 | 11 | 8167 | |||
XM_017017181.1 | XP_016872670.1 | 10 | 8098 | |||
XM_017017182.1 | XP_016872671.1 | 11 | 8167 | |||
XM_017017183.1 | XP_016872672.1 | 10 | 8098 | |||
XM_017017184.1 | XP_016872673.1 | 11 | 8166 | |||
XM_017017185.1 | XP_016872674.1 | 10 | 5994 | |||
XM_017017186.1 | XP_016872675.1 | 10 | 5282 | |||
XM_017017187.1 | XP_016872676.1 | 10 | 9640 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
