siRNA Details
Gene Symbol: TMEM239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318207.1 | NP_001305136.1 | 2 | 3527 | ||
GenBank mRNA | AK126033.1 | - | 2 | 3528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
