siRNA Details
Gene Symbol: MPEG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039396.1 | NP_001034485.1 | 1 | 345 | ||
GenBank mRNA | AK074166.1 | - | 1 | 275 | ||
AK097881.1 | - | 1 | 242 | |||
BC104997.1 | - | 1 | 210 | 8144000 | ||
BC112230.1 | - | 1 | 210 | 8327698 | ||
DA946058.1 | - | 1 | 345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
