siRNA Details
Gene Symbol: MPEG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039396.1 | NP_001034485.1 | 1 | 899 | ||
GenBank mRNA | AK074166.1 | - | 1 | 829 | ||
AK097881.1 | - | 1 | 796 | |||
BC104997.1 | - | 1 | 764 | 8144000 | ||
BC112230.1 | - | 1 | 764 | 8327698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
