siRNA Details
Gene Symbol: RFWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018124.3 | NP_060594.3 | 2 | 418 | ||
XM_005256021.4 | XP_005256078.1 | 3 | 551 | |||
XM_005256022.4 | XP_005256079.1 | 2 | 1301 | |||
XM_006721228.3 | XP_006721291.1 | 3 | 551 | |||
XM_011523191.2 | XP_011521493.1 | 2 | 371 | |||
XM_017023391.1 | XP_016878880.1 | 3 | 467 | |||
XM_017023392.1 | XP_016878881.1 | 2 | 415 | |||
GenBank mRNA | AK291200.1 | - | 3 | 518 | ||
AK296727.1 | - | 2 | 444 | |||
BC059371.1 | - | 2 | 367 | 5266451 | ||
BP872110.1 | - | 2 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
