siRNA Details
Gene Symbol: RFWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018124.3 | NP_060594.3 | 8 | 1313 | ||
XM_005256021.4 | XP_005256078.1 | 9 | 1446 | |||
XM_005256022.4 | XP_005256079.1 | 8 | 2196 | |||
XM_006721228.3 | XP_006721291.1 | 9 | 1446 | |||
XM_011523191.2 | XP_011521493.1 | 8 | 1266 | |||
XM_017023391.1 | XP_016878880.1 | 9 | 1362 | |||
XM_017023392.1 | XP_016878881.1 | 8 | 1310 | |||
GenBank mRNA | AK001382.1 | - | 3 | 215 | ||
AK291200.1 | - | 9 | 1413 | |||
AK315786.1 | - | 5 | 457 | |||
BC002574.2 | - | 6 | 690 | |||
BC059371.1 | - | 8 | 1262 | 5266451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
