siRNA Details
Gene Symbol: RFWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018124.3 | NP_060594.3 | 10 | 1756 | ||
XM_005256021.4 | XP_005256078.1 | 11 | 1889 | |||
XM_005256022.4 | XP_005256079.1 | 10 | 2639 | |||
XM_011523191.2 | XP_011521493.1 | 10 | 1709 | |||
XM_017023391.1 | XP_016878880.1 | 11 | 1805 | |||
GenBank mRNA | AK291200.1 | - | 11 | 1856 | ||
AK315786.1 | - | 7 | 900 | |||
BC002574.2 | - | 8 | 1133 | |||
BC059371.1 | - | 10 | 1705 | 5266451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
