siRNA Details
Gene Symbol: GNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005275.3 | NP_005266.2 | 12 | 6794 | ||
XM_005249015.3 | XP_005249072.1 | 13 | 6257 | |||
GenBank mRNA | AK095071.1 | - | 1 | 1989 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
