siRNA Details
Gene Symbol: KIRREL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286349.1 | NP_001273278.1 | 13 | 3917 | ||
NM_018240.6 | NP_060710.3 | 15 | 4217 | |||
XM_005245305.4 | XP_005245362.1 | 15 | 3922 | |||
GenBank mRNA | AK090554.1 | - | 1 | 600 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
