siRNA Details
Gene Symbol: DUSP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080426.1 | NP_001073895.1 | 5 | 3882 | ||
XM_005245603.3 | XP_005245660.1 | 6 | 4045 | |||
XM_011510146.2 | XP_011508448.1 | 5 | 3919 | |||
GenBank mRNA | AF119045.1 | - | 1 | 2392 | ||
AK094814.1 | - | 1 | 2479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
