siRNA Details
Gene Symbol: DENND4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014856.2 | NP_055671.2 | 24 | 4317 | ||
XM_005245678.4 | XP_005245735.1 | 24 | 4474 | |||
XM_005245679.2 | XP_005245736.1 | 24 | 4324 | |||
XM_011510219.1 | XP_011508521.1 | 22 | 3723 | |||
XM_011510220.2 | XP_011508522.1 | 23 | 4137 | |||
XM_011510221.2 | XP_011508523.1 | 22 | 3845 | |||
XM_017002993.1 | XP_016858482.1 | 21 | 3699 | |||
GenBank mRNA | AB007945.2 | - | 24 | 4317 | ||
AK124049.1 | - | 1 | 982 | |||
AK226053.1 | - | 19 | 2972 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
