siRNA Details
Gene Symbol: DENND4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014856.2 | NP_055671.2 | 4 | 992 | ||
XM_005245678.4 | XP_005245735.1 | 4 | 1149 | |||
XM_005245679.2 | XP_005245736.1 | 4 | 999 | |||
XM_011510219.1 | XP_011508521.1 | 3 | 614 | |||
XM_011510220.2 | XP_011508522.1 | 4 | 982 | |||
XM_011510221.2 | XP_011508523.1 | 4 | 981 | |||
XM_011510222.1 | XP_011508524.1 | 3 | 614 | |||
GenBank mRNA | AB007945.2 | - | 4 | 992 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
