siRNA Details
Gene Symbol: FAM169A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015566.2 | NP_056381.1 | 13 | 2685 | ||
NR_046462.1 | 12 | 2648 | ||||
XM_005248480.2 | XP_005248537.1 | 13 | 2820 | |||
XM_011543306.2 | XP_011541608.1 | 13 | 2907 | |||
XM_011543307.2 | XP_011541609.1 | 13 | 3336 | |||
XM_017009332.1 | XP_016864821.1 | 11 | 3583 | |||
XM_017009333.1 | XP_016864822.1 | 11 | 2557 | |||
GenBank mRNA | AB020695.1 | - | 13 | 2637 | ||
AK022981.1 | - | 4 | 1641 | |||
BX640902.1 | - | 12 | 2462 | |||
BX647536.1 | - | 12 | 2650 | |||
CR936791.1 | - | 13 | 2687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
