siRNA Details
Gene Symbol: FAM169A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015566.2 | NP_056381.1 | 7 | 846 | ||
NR_046462.1 | 6 | 809 | ||||
XM_005248480.2 | XP_005248537.1 | 7 | 981 | |||
XM_011543306.2 | XP_011541608.1 | 7 | 1068 | |||
XM_011543307.2 | XP_011541609.1 | 7 | 1497 | |||
XM_017009332.1 | XP_016864821.1 | 5 | 1744 | |||
XM_017009333.1 | XP_016864822.1 | 5 | 718 | |||
GenBank mRNA | AB020695.1 | - | 7 | 798 | ||
AK290004.1 | - | 7 | 846 | |||
BX640902.1 | - | 6 | 627 | |||
BX647536.1 | - | 6 | 811 | |||
CR936791.1 | - | 7 | 848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
