siRNA Details
Gene Symbol: ZBTB38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080412.2 | NP_001073881.2 | 8 | 6841 | ||
XM_005247256.4 | XP_005247313.1 | 2 | 7361 | |||
XM_005247257.1 | XP_005247314.1 | 3 | 6030 | |||
XM_005247258.1 | XP_005247315.1 | 4 | 6470 | |||
XM_005247261.3 | XP_005247318.1 | 3 | 6099 | |||
XM_011512607.1 | XP_011510909.1 | 4 | 6117 | |||
XM_011512608.1 | XP_011510910.1 | 4 | 6181 | |||
XM_011512611.1 | XP_011510913.1 | 4 | 6122 | |||
XM_017006050.1 | XP_016861539.1 | 5 | 6426 | |||
XM_017006051.1 | XP_016861540.1 | 5 | 6621 | |||
XM_017006052.1 | XP_016861541.1 | 4 | 6544 | |||
XM_017006053.1 | XP_016861542.1 | 5 | 6240 | |||
XM_017006054.1 | XP_016861543.1 | 5 | 6341 | |||
XM_017006055.1 | XP_016861544.1 | 5 | 6245 | |||
XM_017006056.1 | XP_016861545.1 | 5 | 6369 | |||
XM_017006057.1 | XP_016861546.1 | 3 | 6621 | |||
XM_017006058.1 | XP_016861547.1 | 5 | 6264 | |||
XM_017006059.1 | XP_016861548.1 | 4 | 6242 | |||
XM_017006060.1 | XP_016861549.1 | 5 | 6279 | |||
XM_017006061.1 | XP_016861550.1 | 5 | 6220 | |||
GenBank mRNA | AK055693.1 | - | 1 | 1513 | ||
BC022936.1 | - | 1 | 828 | |||
BC051734.1 | - | 1 | 1166 | |||
BC072415.1 | - | 1 | 1774 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AK142305.
Gene Ontology Categories:
