siRNA Details
Gene Symbol: ZBTB38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080412.2 | NP_001073881.2 | 8 | 5399 | ||
XM_005247256.4 | XP_005247313.1 | 2 | 5919 | |||
XM_005247257.1 | XP_005247314.1 | 3 | 4588 | |||
XM_005247258.1 | XP_005247315.1 | 4 | 5028 | |||
XM_005247261.3 | XP_005247318.1 | 3 | 4657 | |||
XM_011512607.1 | XP_011510909.1 | 4 | 4675 | |||
XM_011512608.1 | XP_011510910.1 | 4 | 4739 | |||
XM_011512611.1 | XP_011510913.1 | 4 | 4680 | |||
XM_017006050.1 | XP_016861539.1 | 5 | 4984 | |||
XM_017006051.1 | XP_016861540.1 | 5 | 5179 | |||
XM_017006052.1 | XP_016861541.1 | 4 | 5102 | |||
XM_017006053.1 | XP_016861542.1 | 5 | 4798 | |||
XM_017006054.1 | XP_016861543.1 | 5 | 4899 | |||
XM_017006055.1 | XP_016861544.1 | 5 | 4803 | |||
XM_017006056.1 | XP_016861545.1 | 5 | 4927 | |||
XM_017006057.1 | XP_016861546.1 | 3 | 5179 | |||
XM_017006058.1 | XP_016861547.1 | 5 | 4822 | |||
XM_017006059.1 | XP_016861548.1 | 4 | 4800 | |||
XM_017006060.1 | XP_016861549.1 | 5 | 4837 | |||
XM_017006061.1 | XP_016861550.1 | 5 | 4778 | |||
GenBank mRNA | AK055693.1 | - | 1 | 71 | ||
BC072415.1 | - | 1 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
