siRNA Details
Gene Symbol: DNAJC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015268.3 | NP_056083.3 | 12 | 1324 | ||
XM_005247245.4 | XP_005247302.1 | 12 | 1506 | |||
GenBank mRNA | AB014578.2 | - | 12 | 1373 | ||
AK098729.1 | - | 3 | 268 | |||
AY779857.1 | - | 12 | 1324 | |||
BC043583.1 | - | 12 | 1324 | |||
BC151246.1 | - | 13 | 1394 | 9007477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
