siRNA Details
Gene Symbol: MFAP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009554.3 | NP_001009554.1 | 2 | 3060 | ||
NM_001301647.1 | NP_001288576.1 | 2 | 3135 | |||
NM_001301648.1 | NP_001288577.1 | 3 | 3322 | |||
NM_021647.7 | NP_067679.6 | 3 | 3566 | |||
XM_005263366.3 | XP_005263423.1 | 4 | 3500 | |||
XM_005263368.3 | XP_005263425.1 | 2 | 4674 | |||
XM_017008864.1 | XP_016864353.1 | 5 | 3444 | |||
XM_017008865.1 | XP_016864354.1 | 5 | 6560 | |||
XM_017008866.1 | XP_016864355.1 | 5 | 3634 | |||
XM_017008867.1 | XP_016864356.1 | 5 | 3440 | |||
XM_017008868.1 | XP_016864357.1 | 4 | 3715 | |||
XM_017008869.1 | XP_016864358.1 | 3 | 6547 | |||
XM_017008870.1 | XP_016864359.1 | 3 | 6507 | |||
XM_017008871.1 | XP_016864360.1 | 3 | 3447 | |||
GenBank mRNA | AB014526.1 | - | 3 | 3414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
