siRNA Details
Gene Symbol: CCDC167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138493.2 | NP_612502.1 | 1, 2 | 113 | ||
GenBank mRNA | BC003515.1 | - | 1, 2 | 58 | ||
BC104835.1 | - | 1, 2 | 109 | 8143838 | ||
BC108655.1 | - | 1, 2 | 86 | 6053765 | ||
DA152306.1 | - | 1, 2 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
