siRNA Details
Gene Symbol: CCDC167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138493.2 | NP_612502.1 | 4 | 474 | ||
GenBank mRNA | AF161383.1 | - | 4 | 473 | ||
BC003515.1 | - | 4 | 419 | |||
BC108655.1 | - | 4 | 447 | 6053765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
